This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02682851_g1
See other PRKRA GE Assays ›
Gene Symbol:
PRKRA
Gene Name:
protein activator of interferon induced protein kinase EIF2AK2
Gene Aliases:
-
Chromosome Location:
Chr.36: 22061226 - 22080324 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02682851_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022414743.1 XP_022270451.1 5 - 6 525 168
XM_022414744.1 XP_022270452.1 6 - 7 635 168
XM_022414745.1 XP_022270453.1 5 - 6 502 168
XM_022414746.1 XP_022270454.1 4 - 5 448 168
XM_545545.6 XP_545545.2 6 - 7 734 168

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0