This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02682044_m1
See other ARL6IP4 GE Assays ›
Gene Symbol:
ARL6IP4
Gene Name:
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases:
-
Chromosome Location:
Chr.26: 6522881 - 6525195 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02682044_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARL6IP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014107937.2 XP_013963412.1 823 152
XM_014107938.2 XP_013963413.1 798 152

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0