This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02678337_gH
See other INTS11 GE Assays ›
Gene Symbol:
INTS11
Gene Name:
integrator complex subunit 11
Gene Aliases:
-
Chromosome Location:
Chr.5: 56485690 - 56498074 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02678337_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: INTS11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003639054.3 XP_003639102.1 1461 69
XM_005620376.2 XP_005620433.1 1461 69
XM_005620377.3 XP_005620434.1 1367 69

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0