This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02676468_g1
See other SCNN1D GE Assays ›
Gene Symbol:
SCNN1D
Gene Name:
sodium channel epithelial 1 subunit delta
Gene Aliases:
-
Chromosome Location:
Chr.5: 56455677 - 56466035 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02676468_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCNN1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005620375.3 XP_005620432.1 2078 122
XM_022418891.1 XP_022274599.1 6050 122
XM_022418892.1 XP_022274600.1 5967 122
XM_022418893.1 XP_022274601.1 1439 122
XM_022418894.1 XP_022274602.1 5030 122
XM_022418895.1 XP_022274603.1 4908 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0