This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02675264_m1
See other ACAA1 GE Assays ›
Gene Symbol:
ACAA1
Gene Name:
acetyl-CoA acyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.23: 7889481 - 7901324 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02675264_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACAA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634249.3 XP_005634306.1 10 - 11 1064 111
XM_022408467.1 XP_022264175.1 9 - 10 1005 111
XM_022408468.1 XP_022264176.1 10 - 11 983 111
XM_534222.6 XP_534222.2 9 - 10 1043 111

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0