This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02665502_m1
See other LOC100684448 GE Assays ›
Gene Symbol:
LOC100684448 DNAJB6
Gene Name:
dnaJ homolog subfamily B member 6-like
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases:
-
-
Chromosome Location:
Chr.12: 49763992 - 49765310 on Build CanFam3.1
Chromosome Location:
Chr.16: 19792957 - 19849299 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02665502_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100684448
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003431657.3 XP_003431705.2 1 390 62
Gene Symbol: DNAJB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_856542.5 XP_861635.1 517 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0