Assay Details
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638021.3 | XP_005638078.1 | 3 - 4 | 455 | 139 | |
XM_005638022.3 | XP_005638079.1 | 2 - 3 | 201 | 139 | ||
XM_014109289.2 | XP_013964764.1 | 3 - 4 | 356 | 139 | ||
XM_014109290.2 | XP_013964765.1 | 3 - 4 | 284 | 139 | ||
XM_022412443.1 | XP_022268151.1 | 3 - 4 | 258 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
