Assay Details
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033512.1 | NP_001028684.1 | 2 - 3 | 313 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033512.1 | NP_001028684.1 | 2 - 3 | 313 | 71 |
Set Membership: |
![]() ![]() ![]() |