This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02660809_gH
See other LOC610380 GE Assays ›
Gene Symbol:
LOC610380
Gene Name:
corticotropin-releasing factor receptor 1
Gene Aliases:
CRHR1
Chromosome Location:
Chr.9: 9708696 - 9763281 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02660809_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC610380
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005624197.3 XP_005624254.1 725 59
XM_014116365.2 XP_013971840.1 939 59
XM_847872.5 XP_852965.1 942 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header