This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02660627_m1
See other C20H19ORF53 GE Assays ›
Gene Symbol:
C20H19orf53 MRI1
Gene Name:
chromosome 20 C19orf53 homolog
methylthioribose-1-phosphate isomerase 1
Gene Aliases:
-
-
Chromosome Location:
Chr.20: 48648620 - 48651741 on Build CanFam3.1
Chromosome Location:
Chr.20: 48648987 - 48658231 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02660627_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C20H19orf53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_848432.5 XP_853525.1 2 - 3 410 91
Gene Symbol: MRI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022406981.1 XP_022262689.1 7 - 8 1221 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0