Assay Details
Gene Symbol: SH3GL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425468.1 | XP_022281176.1 | 5 - 6 | 585 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: SH3GL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425468.1 | XP_022281176.1 | 5 - 6 | 585 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |