Assay Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637169.3 | XP_005637226.1 | 5 - 6 | 733 | 104 | |
XM_022411268.1 | XP_022266976.1 | 5 - 6 | 748 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
