This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659942_g1
See other CEBPZOS GE Assays ›
Gene Symbol:
CEBPZOS
Gene Name:
CEBPZ opposite strand
Gene Aliases:
-
Chromosome Location:
Chr.17: 29560205 - 29577984 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02659942_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPZOS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022404637.1 XP_022260345.1 3 - 4 236 89
XM_022404638.1 XP_022260346.1 3 - 4 225 89
XM_022404639.1 XP_022260347.1 3 - 4 409 89
XM_022404640.1 XP_022260348.1 2 - 3 297 89
XM_022404641.1 XP_022260349.1 2 - 3 298 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0