This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659547_m1
See other LOC484411 GE Assays ›
Gene Symbol:
LOC484411 CARD8
Gene Name:
caspase recruitment domain-containing protein 8-like
caspase recruitment domain family, member 8
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 108026553 - 108058664 on Build CanFam3.1
Chromosome Location:
Chr.1: 107994215 - 108025875 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02659547_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC484411
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005616363.3 XP_005616420.1 2 - 3 127 87
XM_005616365.3 XP_005616422.1 2 - 3 126 87
XM_022424383.1 XP_022280091.1 2 - 3 128 87
Gene Symbol: CARD8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022424373.1 XP_022280081.1 3 - 4 507 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005616363.3, XM_005616365.3, XM_022424373.1, XM_022424383.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0