This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659103_m1
See other MRPL9 GE Assays ›
Gene Symbol:
MRPL9
Gene Name:
mitochondrial ribosomal protein L9
Gene Aliases:
-
Chromosome Location:
Chr.17: 60782975 - 60787313 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02659103_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022404375.1 XP_022260083.1 4 - 5 498 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0