Assay Details
Gene Symbol: ANKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636069.3 | XP_005636126.1 | 1454 | 73 | ||
XM_005636070.3 | XP_005636127.1 | 1488 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
