This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02658323_m1
See other ARPIN GE Assays ›
Gene Symbol:
ARPIN
Gene Name:
actin related protein 2/3 complex inhibitor
Gene Aliases:
C3H15orf38
Chromosome Location:
Chr.3: 52870775 - 52880742 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02658323_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARPIN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252380.1 NP_001239309.1 3 - 4 363 61
XM_005618244.1 XP_005618301.1 3 - 4 674 61
XM_022415725.1 XP_022271433.1 3 - 4 277 61
GenBank mRNA DN434974.1 - 3 - 4 372 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_022415725.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0