Assay Details
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846951.5 | XP_852044.2 | 784 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846951.5 | XP_852044.2 | 784 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() |