This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02657807_m1
See other LOC607830 GE Assays ›
Gene Symbol:
LOC607830 TMEM9
Gene Name:
transmembrane protein 9-like
transmembrane protein 9
Gene Aliases:
-
-
Chromosome Location:
Chr.7: 1705803 - 1723905 on Build CanFam3.1
Chromosome Location:
Chr.7: 2069017 - 2087961 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02657807_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC607830
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014114954.1 XP_013970429.1 269 77
Gene Symbol: TMEM9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003639175.3 XP_003639223.1 359 77
XM_005622265.3 XP_005622322.1 329 77
XM_014115042.2 XP_013970517.1 409 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0