This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02657775_m1
See other MIA2 GE Assays ›
Gene Symbol:
MIA2
Gene Name:
MIA SH3 domain ER export factor 2
Gene Aliases:
CTAGE5
Chromosome Location:
Chr.8: 17390738 - 17482421 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02657775_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014115855.2 XP_013971330.1 19 - 20 3055 144
XM_014115856.2 XP_013971331.1 18 - 19 3045 144
XM_014115857.2 XP_013971332.1 18 - 19 3027 144
XM_014115858.2 XP_013971333.1 17 - 18 3018 144
XM_014115859.2 XP_013971334.1 19 - 20 3056 144
XM_014115860.2 XP_013971335.1 14 - 15 1137 144
XM_014115861.2 XP_013971336.1 14 - 15 1157 144
XM_014115862.2 XP_013971337.1 13 - 14 1130 144
XM_014115863.2 XP_013971338.1 14 - 15 1123 144
XM_014115864.2 XP_013971339.1 14 - 15 1024 144
XM_014115865.2 XP_013971340.1 14 - 15 1175 144
XM_022421957.1 XP_022277665.1 13 - 14 1130 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0