This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02657586_m1
See other UBE2U GE Assays ›
Gene Symbol:
UBE2U
Gene Name:
ubiquitin conjugating enzyme E2 U
Gene Aliases:
-
Chromosome Location:
Chr.5: 45947637 - 45998353 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02657586_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2U
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022418718.1 XP_022274426.1 1 - 2 495 97
XM_022418719.1 XP_022274427.1 1 - 2 499 97
XM_022418720.1 XP_022274428.1 1 - 2 489 97
XM_022418721.1 XP_022274429.1 1 - 2 501 97
XM_022418722.1 XP_022274430.1 1 - 2 501 97
XM_022418724.1 XP_022274432.1 1 - 2 488 97
XM_022418725.1 XP_022274433.1 1 - 2 490 97
XM_022418726.1 XP_022274434.1 1 - 2 842 97
XM_022418727.1 XP_022274435.1 1 - 2 504 97
XM_022418730.1 XP_022274438.1 1 - 2 491 97
XM_022418731.1 XP_022274439.1 1 - 2 491 97
XM_022418732.1 XP_022274440.1 1 - 2 496 97
XM_022418733.1 XP_022274441.1 1 - 2 493 97
XM_022418734.1 XP_022274442.1 1 - 2 492 97
XM_022418735.1 XP_022274443.1 1 - 2 494 97
XM_022418737.1 XP_022274445.1 1 - 2 503 97
XM_022418738.1 XP_022274446.1 1 - 2 495 97
XM_022418740.1 XP_022274448.1 1 - 2 494 97
XM_022418741.1 XP_022274449.1 1 - 2 57 97
XM_022418745.1 XP_022274453.1 1 - 2 486 97
XM_022418746.1 XP_022274454.1 1 - 2 838 97
XM_022418747.1 XP_022274455.1 1 - 2 500 97
XM_022418748.1 XP_022274456.1 1 - 2 839 97
XM_022418750.1 XP_022274458.1 1 - 2 57 97
XM_847284.5 XP_852377.2 1 - 2 485 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_022418741.1, XM_022418750.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0