Assay Details
Gene Symbol: MOSMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845845.4 | XP_850938.1 | 1 - 2 | 121 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: MOSMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845845.4 | XP_850938.1 | 1 - 2 | 121 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |