This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02656890_g1
See other APOOL GE Assays ›
Gene Symbol:
APOOL
Gene Name:
apolipoprotein O like
Gene Aliases:
-
Chromosome Location:
Chr.X: 65850365 - 65953013 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02656890_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOOL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014111952.2 XP_013967427.1 7 - 8 707 68
XM_022415978.1 XP_022271686.1 7 - 8 712 68
XM_022415979.1 XP_022271687.1 6 - 7 610 68
XM_022415980.1 XP_022271688.1 6 - 7 567 68
XM_022415981.1 XP_022271689.1 7 - 8 543 68
XM_845869.5 XP_850962.2 6 - 7 609 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0