This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02656628_m1
See other BABAM2 GE Assays ›
Gene Symbol:
BABAM2
Gene Name:
BRISC and BRCA1 A complex member 2
Gene Aliases:
BRE
Chromosome Location:
Chr.17: 21886127 - 22288513 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02656628_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BABAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005630287.2 XP_005630344.1 6 - 7 739 84
XM_022404584.1 XP_022260292.1 6 - 7 723 84
XM_022404585.1 XP_022260293.1 6 - 7 739 84
XM_022404586.1 XP_022260294.1 6 - 7 739 84
XM_022404587.1 XP_022260295.1 6 - 7 739 84
XM_022404588.1 XP_022260296.1 6 - 7 739 84
XM_845588.4 XP_850681.1 6 - 7 739 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0