This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02655176_m1
See other CCDC187 GE Assays ›
Gene Symbol:
CCDC187
Gene Name:
coiled-coil domain containing 187
Gene Aliases:
-
Chromosome Location:
Chr.9: 49165109 - 49202935 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02655176_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCDC187
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022423591.1 XP_022279299.1 9 - 10 3019 89
XM_022423592.1 XP_022279300.1 8 - 9 2807 89
XM_022423593.1 XP_022279301.1 8 - 9 2807 89
XM_022423594.1 XP_022279302.1 8 - 9 2807 89
XM_022423595.1 XP_022279303.1 8 - 9 3129 89
XM_022423596.1 XP_022279304.1 8 - 9 2812 89
XM_022423597.1 XP_022279305.1 7 - 8 2675 89
XM_022423598.1 XP_022279306.1 8 - 9 2807 89
XM_022423599.1 XP_022279307.1 4 - 5 1990 89
XM_022423600.1 XP_022279308.1 4 - 5 2010 89
XM_022423601.1 XP_022279309.1 8 - 9 2807 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein non-motor microtubule binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0