This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02654715_m1
See other LOC111091303 GE Assays ›
Gene Symbol:
LOC111091303 RPP14
Gene Name:
hydroxyacyl-thioester dehydratase type 2, mitochondrial-like
ribonuclease P/MRP subunit p14
Gene Aliases:
-
-
Chromosome Location:
Chr.20: 32280815 - 32288844 on Build CanFam3.1
Chromosome Location:
Chr.20: 32282225 - 32290576 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02654715_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC111091303
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022406528.1 XP_022262236.1 1 - 2 75 84
Gene Symbol: RPP14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_844095.5 XP_849188.2 3 - 4 300 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_022406528.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0