Assay Details
Gene Symbol: EGFL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022415908.1 | XP_022271616.1 | 13 - 14 | 1934 | 59 | |
XM_548865.6 | XP_548865.3 | 8 - 9 | 1212 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
