This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02653530_m1
See other HSD17B3 GE Assays ›
Gene Symbol:
HSD17B3
Gene Name:
hydroxysteroid 17-beta dehydrogenase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 70551013 - 70586364 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02653530_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD17B3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003638870.4 XP_003638918.1 8 - 9 642 65
XM_005615786.3 XP_005615843.1 7 - 8 606 65
XM_005615787.3 XP_005615844.1 9 - 10 711 65
XM_022420319.1 XP_022276027.1 8 - 9 642 65
XM_022420325.1 XP_022276033.1 7 - 8 516 65
XM_022420326.1 XP_022276034.1 6 - 7 441 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0