This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02653521_m1
See other ILDR2 GE Assays ›
Gene Symbol:
ILDR2
Gene Name:
immunoglobulin like domain containing receptor 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 31409015 - 31469741 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02653521_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ILDR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005622565.3 XP_005622622.1 7 - 8 1150 68
XM_022421114.1 XP_022276822.1 7 - 8 1149 68
XM_022421115.1 XP_022276823.1 6 - 7 1093 68
XM_022421116.1 XP_022276824.1 6 - 7 983 68
XM_022421117.1 XP_022276825.1 4 - 5 781 68
XM_547485.6 XP_547485.3 7 - 8 1177 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0