Assay Details
Gene Symbol: DAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414350.1 | XP_022270058.1 | 114 | 98 | ||
XM_022414351.1 | XP_022270059.1 | 114 | 98 | |||
XM_843925.5 | XP_849018.1 | 114 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
