This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02652741_m1
See other RCAN3 GE Assays ›
Gene Symbol:
RCAN3
Gene Name:
RCAN family member 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 75118201 - 75167309 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02652741_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RCAN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005617795.3 XP_005617852.1 3 - 4 565 63
XM_005617796.3 XP_005617853.1 3 - 4 565 63
XM_005617797.3 XP_005617854.1 3 - 4 565 63
XM_005617798.3 XP_005617855.1 3 - 4 531 63
XM_014110572.2 XP_013966047.1 2 - 3 389 63
XM_022413261.1 XP_022268969.1 3 - 4 565 63
XM_022413266.1 XP_022268974.1 3 - 4 565 63
XM_022413267.1 XP_022268975.1 3 - 4 565 63
XM_544495.5 XP_544495.2 3 - 4 616 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005617798.3, XM_014110572.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header