This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02652706_m1
See other POLR1H GE Assays ›
Gene Symbol:
POLR1H
Gene Name:
RNA polymerase I subunit H
Gene Aliases:
ZNRD1
Chromosome Location:
Chr.35: 26285520 - 26288550 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02652706_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR1H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005640147.3 XP_005640204.1 1 - 2 414 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0