This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02652435_m1
See other IGFBP2 GE Assays ›
Gene Symbol:
IGFBP2
Gene Name:
insulin like growth factor binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.37: 23553227 - 23569321 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02652435_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022415094.1 XP_022270802.1 2 - 3 327 96
XM_022415095.1 XP_022270803.1 2 - 3 270 96
XM_022415096.1 XP_022270804.1 2 - 3 267 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0