This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02650370_m1
See other ERP27 GE Assays ›
Gene Symbol:
ERP27
Gene Name:
endoplasmic reticulum protein 27
Gene Aliases:
-
Chromosome Location:
Chr.27: 31682586 - 31702959 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02650370_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERP27
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022411255.1 XP_022266963.1 1 - 2 433 62
XM_543794.6 XP_543794.3 1 - 2 319 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header