Assay Details
Gene Symbol: POGLUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022418348.1 | XP_022274056.1 | 6 - 7 | 1141 | 139 | |
XM_546537.4 | XP_546537.2 | 5 - 6 | 1108 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
