Assay Details
Gene Symbol: BOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639493.3 | XP_005639550.1 | 15 - 16 | 2892 | 61 | |
XM_014110260.2 | XP_013965735.1 | 17 - 18 | 3525 | 61 | ||
XM_014110261.2 | XP_013965736.1 | 18 - 19 | 3645 | 61 | ||
XM_014110263.2 | XP_013965738.1 | 16 - 17 | 3266 | 61 | ||
XM_014110264.2 | XP_013965739.1 | 15 - 16 | 2764 | 61 | ||
XM_022413967.1 | XP_022269675.1 | 18 - 19 | 3650 | 61 | ||
XM_022413968.1 | XP_022269676.1 | 19 - 20 | 3641 | 61 | ||
XM_022413969.1 | XP_022269677.1 | 18 - 19 | 3516 | 61 | ||
XM_022413970.1 | XP_022269678.1 | 17 - 18 | 3165 | 61 | ||
XM_022413971.1 | XP_022269679.1 | 17 - 18 | 3543 | 61 | ||
XM_022413972.1 | XP_022269680.1 | 17 - 18 | 3391 | 61 | ||
XM_022413973.1 | XP_022269681.1 | 16 - 17 | 2908 | 61 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
