This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02649873_m1
See other SEL1L2 GE Assays ›
Gene Symbol:
SEL1L2
Gene Name:
SEL1L2 adaptor subunit of ERAD E3 ligase
Gene Aliases:
-
Chromosome Location:
Chr.24: 8678528 - 8812373 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02649873_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEL1L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634769.3 XP_005634826.1 12 - 13 1220 88
XM_005634770.3 XP_005634827.1 12 - 13 1219 88
XM_005634772.2 XP_005634829.1 10 - 11 970 88
XM_014107052.2 XP_013962527.1 12 - 13 1226 88
XM_014107053.2 XP_013962528.1 12 - 13 1532 88
XM_022408898.1 XP_022264606.1 12 - 13 1219 88
XM_022408900.1 XP_022264608.1 8 - 9 922 88
XM_022408901.1 XP_022264609.1 9 - 10 807 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0