Assay Details
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639662.4 | XP_003639710.4 | 4 - 5 | 672 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639662.4 | XP_003639710.4 | 4 - 5 | 672 | 90 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |