This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02648653_m1
See other GPRC5C GE Assays ›
Gene Symbol:
GPRC5C
Gene Name:
G protein-coupled receptor class C group 5 member C
Gene Aliases:
-
Chromosome Location:
Chr.9: 5983545 - 6005586 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02648653_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC5C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014116321.2 XP_013971796.1 1500 80
XM_014116322.2 XP_013971797.1 1421 80
XM_014116323.1 XP_013971798.1 1985 80
XM_014116324.2 XP_013971799.1 1185 80
XM_014116325.2 XP_013971800.1 1226 80
XM_014116326.2 XP_013971801.1 1220 80
XM_014116327.2 XP_013971802.1 1498 80
XM_014116328.2 XP_013971803.1 1300 80
XM_014116329.2 XP_013971804.1 1299 80
XM_022422747.1 XP_022278455.1 1501 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0