This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02647306_g1
See other MUTYH GE Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.15: 15018345 - 15026539 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02647306_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MUTYH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014119532.2 XP_013975007.1 2 - 3 365 59
XM_014119533.2 XP_013975008.1 2 - 3 93 59
XM_014119534.2 XP_013975009.1 2 - 3 282 59
XM_014119535.2 XP_013975010.1 2 - 3 178 59
XM_022428007.1 XP_022283715.1 2 - 3 178 59
XM_022428008.1 XP_022283716.1 2 - 3 177 59
XM_022428009.1 XP_022283717.1 2 - 3 177 59
XM_022428010.1 XP_022283718.1 2 - 3 178 59
XM_022428011.1 XP_022283719.1 2 - 3 371 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014119533.2, XM_014119534.2, XM_014119535.2, XM_022428007.1, XM_022428008.1, XM_022428009.1, XM_022428010.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0