This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02647139_m1
See other NUDT6 GE Assays ›
Gene Symbol:
NUDT6
Gene Name:
nudix hydrolase 6
Gene Aliases:
-
Chromosome Location:
Chr.19: 17371589 - 17403182 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02647139_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014121452.2 XP_013976927.1 577 119
XM_022405963.1 XP_022261671.1 289 119

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_022405963.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

nucleotide phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0