Assay Details
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536768.6 | XP_536768.2 | 3 - 4 | 360 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536768.6 | XP_536768.2 | 3 - 4 | 360 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() |