Assay Details
Gene Symbol: PYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538222.6 | XP_538222.2 | 586 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: PYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538222.6 | XP_538222.2 | 586 | 90 |
Set Membership: |
![]() ![]() ![]() |