This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02645440_m1
See other PRXL2A GE Assays ›
Gene Symbol:
PRXL2A
Gene Name:
peroxiredoxin like 2A
Gene Aliases:
C4H10orf58, FAM213A
Chromosome Location:
Chr.4: 29539182 - 29559353 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02645440_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRXL2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619093.3 XP_005619150.3 3 - 4 434 67
XM_005619094.1 XP_005619151.1 4 - 5 454 67
XM_022417605.1 XP_022273313.1 3 - 4 313 67
XM_536403.7 XP_536403.3 3 - 4 393 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0