This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02644755_m1
See other COP1 GE Assays ›
Gene Symbol:
COP1
Gene Name:
COP1 E3 ubiquitin ligase
Gene Aliases:
RFWD2
Chromosome Location:
Chr.7: 23320483 - 23565093 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02644755_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014115170.2 XP_013970645.2 8 - 9 671 73
XM_014115171.2 XP_013970646.1 7 - 8 983 73
XM_014115172.2 XP_013970647.1 9 - 10 1217 73
XM_022421014.1 XP_022276722.1 7 - 8 992 73
XM_022421015.1 XP_022276723.1 7 - 8 980 73
XM_022421016.1 XP_022276724.1 9 - 10 1082 73
XM_537181.6 XP_537181.2 8 - 9 1051 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0