This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02643844_m1
See other RAMAC GE Assays ›
Gene Symbol:
RAMAC
Gene Name:
RNA guanine-7 methyltransferase activating subunit
Gene Aliases:
FAM103A1, RAMMET
Chromosome Location:
Chr.3: 54776831 - 54782047 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02643844_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAMAC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_536205.6 XP_536205.1 2 - 3 74 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_536205.6 transcripts.

Set Membership:

Amplicon length between 86 and 100 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header