This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02642347_m1
See other SUPT3H GE Assays ›
Gene Symbol:
SUPT3H
Gene Name:
SPT3 homolog, SAGA and STAGA complex component
Gene Aliases:
-
Chromosome Location:
Chr.12: 13083084 - 13684090 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02642347_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUPT3H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022426111.1 XP_022281819.1 8 - 9 996 104
XM_022426113.1 XP_022281821.1 8 - 9 784 104
XM_022426114.1 XP_022281822.1 8 - 9 762 104
XM_022426115.1 XP_022281823.1 8 - 9 994 104
XM_022426116.1 XP_022281824.1 8 - 9 818 104
XM_022426117.1 XP_022281825.1 8 - 9 780 104
XM_022426118.1 XP_022281826.1 7 - 8 609 104
XM_532157.7 XP_532157.5 8 - 9 994 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0