This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02639691_g1
See other LOC100682812 GE Assays ›
Gene Symbol:
LOC100682812 MRPL42
Gene Name:
39S ribosomal protein L42, mitochondrial-like
mitochondrial ribosomal protein L42
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 30064401 - 30064965 on Build CanFam3.1
Chromosome Location:
Chr.15: 33701938 - 33732885 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02639691_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100682812
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014112267.2 XP_013967742.1 4 - 5 433 154
Gene Symbol: MRPL42
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022403311.1 XP_022259019.1 6 - 7 600 150
XM_022403312.1 XP_022259020.1 6 - 7 621 150
XM_022403313.1 XP_022259021.1 6 - 7 637 150
XM_022403314.1 XP_022259022.1 6 - 7 511 150
XM_532651.6 XP_532651.1 5 - 6 579 150

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0