This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02639553_m1
See other FAM71F1 GE Assays ›
Gene Symbol:
FAM71F1
Gene Name:
family with sequence similarity 71 member F1
Gene Aliases:
-
Chromosome Location:
Chr.14: 7905747 - 7925117 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02639553_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM71F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005628416.3 XP_005628473.1 2 - 3 548 82
XM_014118882.2 XP_013974357.1 3 - 4 723 82
XM_014118883.2 XP_013974358.1 3 - 4 723 82
XM_022427258.1 XP_022282966.1 3 - 4 723 82
XM_022427259.1 XP_022282967.1 3 - 4 720 82
XM_022427260.1 XP_022282968.1 3 - 4 720 82
XM_022427261.1 XP_022282969.1 3 - 4 720 82
XM_022427262.1 XP_022282970.1 3 - 4 471 82
XM_022427263.1 XP_022282971.1 3 - 4 430 82
XM_022427265.1 XP_022282973.1 3 - 4 723 82
XM_022427266.1 XP_022282974.1 2 - 3 332 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header