This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02638220_g1
See other CD209 GE Assays ›
Gene Symbol:
CD209
Gene Name:
CD209 molecule
Gene Aliases:
CLEC4M
Chromosome Location:
Chr.20: 52517759 - 52542085 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02638220_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD209
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130832.1 NP_001124304.1 6 - 7 481 75
XM_005632335.3 XP_005632392.1 6 - 7 565 75
XM_005632336.3 XP_005632393.1 5 - 6 487 75
XM_022406247.1 XP_022261955.1 6 - 7 890 75
GenBank mRNA EU670674.1 - 6 - 7 481 75
EU670675.1 - 5 - 6 442 75
EU670676.1 - 5 - 6 418 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0